Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010879.3 | NP_035009.3 | 5 | 1827 | ||
GenBank mRNA | AK014949.1 | - | 5 | 1821 | ||
AK051665.1 | - | 5 | 1827 | |||
BC011071.1 | - | 3 | 1392 | 3673483 | ||
BC034255.1 | - | 5 | 1567 | 3157491 |