Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011020.3 | NP_035150.3 | 2, 3 | 227 | ||
GenBank mRNA | AB001926.1 | - | 2, 3 | 193 | ||
AK032066.1 | - | 2, 3 | 257 | |||
AK144225.1 | - | 2, 3 | 264 | |||
AK163459.1 | - | 3, 4 | 399 | |||
BC012712.1 | - | 2, 3 | 296 | 4190161 | ||
BC057002.1 | - | 2, 3 | 227 | 5702602 | ||
BC110662.1 | - | 2, 3 | 247 | 4511154 | ||
D49482.1 | - | 2, 3 | 221 | |||
U23921.1 | - | 2, 3 | 344 |