Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011034.4 | NP_035164.1 | 4 | 376 | ||
GenBank mRNA | AK002287.1 | - | 4 | 375 | ||
AK008711.1 | - | 4 | 378 | |||
AK010688.1 | - | 4 | 377 | |||
AK083243.1 | - | 4 | 377 | |||
AK145138.1 | - | 377 | ||||
AK150797.1 | - | 4 | 377 | |||
AK151459.1 | - | 4 | 377 | |||
AK163984.1 | - | 4 | 377 | |||
AK167624.1 | - | 4 | 377 | |||
AK169154.1 | - | 4 | 374 | |||
BC083348.1 | - | 3 | 328 | 5710010 | ||
BC086648.1 | - | 4 | 342 | 6819164 | ||
D16142.1 | - | 4 | 362 | |||
D21252.1 | - | 4 | 366 |