Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011034.4 | NP_035164.1 | 6 | 1055 | ||
GenBank mRNA | AK083243.1 | - | 6 | 1056 | ||
AK145138.1 | - | 1054 | ||||
AK167624.1 | - | 6 | 1056 |