Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013566.2 | NP_038594.2 | 6, 7 | 1061 | ||
GenBank mRNA | AK078871.1 | - | 6, 7 | 1062 | ||
AK144773.1 | - | 6, 7 | 1033 | |||
AK154766.1 | - | 6, 7 | 1062 | |||
AK155207.1 | - | 6, 7 | 1028 | |||
AK155594.1 | - | 6, 7 | 1034 | |||
BC011184.1 | - | 6, 7 | 1034 | 4187781 | ||
M68903.1 | - | 6, 7 | 1009 | |||
M95632.1 | - | 6, 7 | 1005 | |||
M95633.1 | - | 6, 7 | 1005 | |||
S44607.1 | - | 5, 6 | 973 |