Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Madcam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013591.2 | NP_038619.2 | 5 | 1253 | ||
GenBank mRNA | AK137028.1 | - | 5 | 1254 | ||
AK156096.1 | - | 4 | 799 | |||
BC024372.1 | - | 4 | 814 | 4986802 | ||
BC115910.1 | - | 4 | 815 | 40105157 | ||
BC118056.1 | - | 4 | 815 | 40105156 | ||
L21203.1 | - | 5 | 1250 |