Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112698.2 | NP_001106168.1 | 2 | 147 | ||
NM_013609.3 | NP_038637.1 | 3 | 274 | |||
GenBank mRNA | AK144588.1 | - | 3 | 266 | ||
BC011123.1 | - | 2 | 240 | |||
BF786523.1 | - | 2 | 161 | |||
K01759.1 | - | 2 | 241 | |||
M14805.1 | - | 2 | 241 | |||
M35075.1 | - | 3 | 262 | |||
S62089.1 | - | 1 | 121 | |||
V00836.1 | - | 3 | 262 |