Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KTN1 SIRNA Assays ›
Gene Symbol:
KTN1
Gene Name:
kinectin 1
Gene Aliases:
CG1, KNT, MU-RMS-40.19
Chromosome Location:
Chr.14: 55580207 - 55684584 on Build GRCh38
UniGene ID:
Hs.509414
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
15686
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KTN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079521.1
NP_001072989.1
44
4286
NM_001079522.1
NP_001072990.1
43
4257
NM_001271014.1
NP_001257943.1
44
4326
NM_004986.3
NP_004977.2
42
4115
NR_073128.1
44
4313
NR_073129.1
46
4466
XM_006720138.2
XP_006720201.1
45
4401
XM_006720139.2
XP_006720202.1
43
4165
XM_006720140.3
XP_006720203.1
43
4162
XM_006720141.2
XP_006720204.1
42
4072
XM_011536751.2
XP_011535053.1
44
4507
XM_011536752.1
XP_011535054.1
43
4180
XM_011536753.1
XP_011535055.1
42
4096
XM_017021278.1
XP_016876767.1
44
10099
XM_017021279.1
XP_016876768.1
42
4093
XM_017021280.1
XP_016876769.1
43
4245
XM_017021281.1
XP_016876770.1
43
10067
XM_017021282.1
XP_016876771.1
43
4233
XM_017021283.1
XP_016876772.1
43
4167
XM_017021284.1
XP_016876773.1
41
4009
XM_017021285.1
XP_016876774.1
42
4141
XM_017021286.1
XP_016876775.1
40
3919
GenBank mRNA
AJ270687.1
-
1
57
AK055870.1
-
9
2039
AK124180.1
-
4
1987
AK125499.1
-
11
1546
AK300703.1
-
12
1273
AK302797.1
-
44
4280
AK308272.1
-
25
1935
AY264265.1
-
46
4351
AY536375.1
-
34
2499
BC117132.1
-
42
4074
40125683
BC143720.1
-
43
4143
9052234
BG206718.1
-
2
205
BX537639.1
-
4
4857
D13629.1
-
42
4020
L25616.1
-
41
3977
Z22551.1
-
44
4174
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule-based movement
cell-cell adhesion
protein binding
kinesin binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0