Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MECP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004992.3 | NP_004983.1 | 4 | 3951 | ||
XM_005274682.4 | XP_005274739.1 | 5 | 4025 | |||
XM_006724819.3 | XP_006724882.1 | 4 | 3702 | |||
XM_011531166.2 | XP_011529468.1 | 3 | 3838 | |||
XM_017029533.1 | XP_016885022.1 | 7 | 4451 | |||
GenBank mRNA | AB209464.1 | - | 3 | 2657 | ||
AF158180.1 | - | 4 | 3892 | |||
AJ132917.1 | - | 3 | 3800 | |||
GQ203293.1 | - | 4 | 3952 | |||
GQ203294.1 | - | 4 | 3951 | |||
GQ203295.1 | - | 4 | 3953 | |||
GQ896382.1 | - | 4 | 3951 | |||
GU479943.1 | - | 4 | 3951 | |||
HM020402.1 | - | 4 | 3947 | |||
HM156732.1 | - | 4 | 3951 | |||
HM156733.1 | - | 4 | 3952 | |||
HQ141378.1 | - | 4 | 3953 | |||
HQ154629.1 | - | 5 | 3611 |