Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164190.1 | NP_001157662.1 | 9 | 1021 | ||
NM_001164191.1 | NP_001157663.1 | 9 | 912 | |||
NM_001164192.1 | NP_001157664.1 | 9 | 912 | |||
NM_019926.3 | NP_064310.2 | 9 | 901 | |||
GenBank mRNA | AF073996.1 | - | 9 | 902 | ||
AK016445.1 | - | 9 | 1022 | |||
AK031794.1 | - | 9 | 883 | |||
AK166452.1 | - | 9 | 880 | |||
AK171475.1 | - | 9 | 891 | |||
AK220524.1 | - | 3022 | ||||
BC051022.1 | - | 9 | 870 | |||
BC066847.1 | - | 9 | 859 | |||
BC090984.1 | - | 9 | 879 | 30246206 |