Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025922.2 | NP_080198.2 | 6, 7 | 444 | ||
GenBank mRNA | AB100501.1 | - | 6, 7 | 456 | ||
AK008279.1 | - | 6, 7 | 484 | |||
AK009248.1 | - | 6, 7 | 456 | |||
AK138101.1 | - | 6, 7 | 689 | |||
AK153904.1 | - | 6, 7 | 463 | |||
BC026508.1 | - | 6, 7 | 444 | 4512398 | ||
BC094466.1 | - | 6, 7 | 432 | 4936811 |