Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031170.2 | NP_112447.2 | 8 | 1358 | ||
GenBank mRNA | AK077597.1 | - | 8 | 1331 | ||
AK145546.1 | - | 8 | 1331 | |||
AK145679.1 | - | 8 | 1331 | |||
AK145777.1 | - | 8 | 1358 | |||
AK146948.1 | - | 8 | 1328 | |||
AK147044.1 | - | 8 | 1328 | |||
AK166737.1 | - | 8 | 1331 | |||
AK166768.1 | - | 8 | 1328 | |||
AK166854.1 | - | 8 | 1331 | |||
AK166960.1 | - | 8 | 1331 | |||
AK166993.1 | - | 8 | 1353 | |||
AK167120.1 | - | 8 | 1328 | |||
AK167269.1 | - | 8 | 1331 | |||
AK167471.1 | - | 8 | 1328 | |||
AK168522.1 | - | 8 | 1328 | |||
AK168540.1 | - | 8 | 1328 | |||
AK168561.1 | - | 8 | 1328 | |||
AK168726.1 | - | 8 | 1328 | |||
AK168910.1 | - | 8 | 1328 | |||
AK169136.1 | - | 8 | 1331 | |||
AK169691.1 | - | 8 | 1329 | |||
BC094009.1 | - | 8 | 1322 | 4987971 | ||
BC106154.1 | - | 8 | 1309 | 3994432 | ||
M21836.1 | - | 8 | 1319 | |||
M22831.1 | - | 8 | 1290 | |||
X12789.1 | - | 8 | 1628 |