Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032394.3 | NP_115770.2 | 8 | 834 | ||
GenBank mRNA | AF242411.1 | - | 8 | 871 | ||
BC138338.1 | - | 8 | 834 | 8861358 | ||
BC138341.1 | - | 8 | 834 | 8861361 |