Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ednrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136061.2 | NP_001129533.1 | 2 | 781 | ||
NM_001276296.1 | NP_001263225.1 | 1 | 595 | |||
NM_007904.4 | NP_031930.1 | 1 | 595 | |||
GenBank mRNA | AK076426.1 | - | 1 | 589 | ||
AK082103.1 | - | 2 | 582 | |||
AK083415.1 | - | 2 | 478 | |||
AK085165.1 | - | 1 | 586 | |||
AK085532.1 | - | 1 | 591 | |||
AK192059.1 | - | 1 | 48 | |||
AK195461.1 | - | 1 | 44 | |||
AK202614.1 | - | 1 | 126 | |||
AK211790.1 | - | 1 | 46 | |||
AK219164.1 | - | 1 | 39 | |||
BC026553.1 | - | 2 | 707 | 4971909 | ||
BQ442271.1 | - | 1 | 596 | |||
U32329.1 | - | 1 | 573 |