Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207053.1 | NP_001193982.1 | 2 | 227 | ||
NM_005046.3 | NP_005037.1 | 3 | 358 | |||
NM_139277.2 | NP_644806.1 | 3 | 188 | |||
GenBank mRNA | AA101043.1 | - | 3 | 188 | ||
AF411214.1 | - | 3 | 332 | |||
AF411215.1 | - | 2 | 201 | |||
AK289660.1 | - | 3 | 211 | |||
AY601109.1 | - | 2 | 86 | |||
BC032005.1 | - | 3 | 197 | 4750452 | ||
DQ018784.1 | - | 2 | 86 | |||
KF963189.1 | - | 2 | 50 | |||
KF963190.1 | - | 2 | 51 | |||
KF963191.1 | - | 2 | 51 | |||
KF963192.1 | - | 2, 3 | 50 | |||
KF963193.1 | - | 2 | 50 | |||
L33404.1 | - | 2 | 101 |