Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163131.1 | NP_001156603.1 | 9 | 1970 | ||
NM_007921.3 | NP_031947.1 | 9 | 1910 | |||
GenBank mRNA | AF016294.1 | - | 9 | 1876 | ||
AK143135.1 | - | 9 | 1960 |