Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007927.3 | NP_031953.1 | 2 | 369 | ||
GenBank mRNA | AK146361.1 | - | 2 | 215 | ||
AK160531.1 | - | 2 | 214 | |||
AK167852.1 | - | 2 | 212 | |||
AK188967.1 | - | 2 | 68 | |||
BC132131.1 | - | 2 | 181 | 40130408 | ||
BC132133.1 | - | 2 | 181 | 40130410 | ||
BY260524.1 | - | 2 | 370 | |||
U73902.1 | - | 2 | 219 |