Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007937.3 | NP_031963.2 | 1, 2 | 1064 | ||
GenBank mRNA | AK034289.1 | - | 1, 2 | 1063 | ||
AK049740.1 | - | 1, 2 | 1065 | |||
AK089130.1 | - | 1, 2 | 459 | |||
AK089143.1 | - | 1, 2 | 923 | |||
AK133698.1 | - | 1, 2 | 1064 | |||
BC057401.1 | - | 1, 2 | 1042 | 5697758 | ||
U07357.1 | - | 1, 2 | 604 |