Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fasn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007988.3 | NP_032014.3 | 15 | 2895 | ||
GenBank mRNA | AF127033.1 | - | 15 | 2412 | ||
AK147214.1 | - | 15 | 2591 | |||
AK147694.1 | - | 15 | 2658 | |||
AK171499.1 | - | 15 | 2527 | |||
BC046513.1 | - | 15 | 2449 | 4196969 |