Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014602.2 | NP_055417.1 | 2 | 805 | ||
GenBank mRNA | AK314219.1 | - | 2 | 752 | ||
BC017332.2 | - | 2 | 770 | |||
BC057239.1 | - | 2 | 786 | |||
BC110318.1 | - | 2 | 751 | 5492292 | ||
BC127105.1 | - | 2 | 622 | 40124707 | ||
BC127106.1 | - | 2 | 622 | 40124708 | ||
Y08991.1 | - | 2 | 791 |