Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301412.1 | NP_001288341.1 | 4, 5 | 694 | ||
NM_008017.4 | NP_032043.3 | 4, 5 | 618 | |||
GenBank mRNA | AJ534939.1 | - | 3, 4 | 449 | ||
AK013109.1 | - | 4, 5 | 660 | |||
AK017305.1 | - | 4, 5 | 663 | |||
AK017747.1 | - | 4, 5 | 597 | |||
AK028233.1 | - | 4, 5 | 660 | |||
AK145341.1 | - | 4, 5 | 648 | |||
AK152090.1 | - | 4, 5 | 647 | |||
AK161661.1 | - | 4, 5 | 588 | |||
BC011064.1 | - | 4, 5 | 556 | |||
BC021847.1 | - | 4, 5 | 706 | |||
BC094380.1 | - | 4, 5 | 590 | 30543190 | ||
CJ049961.1 | - | 4, 5 | 619 |