Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122892.1 | NP_001116364.1 | 10 | 1523 | ||
NM_001122893.1 | NP_001116365.1 | 11 | 1602 | |||
NM_008054.2 | NP_032080.2 | 11 | 1593 | |||
GenBank mRNA | AK156584.1 | - | 11 | 1604 | ||
AK171646.1 | - | 11 | 1595 | |||
BC010554.1 | - | 10 | 1204 | |||
BC032149.1 | - | 10 | 1238 | 3602546 | ||
BC092217.1 | - | 10 | 1519 | 6403711 | ||
M27266.1 | - | 12 | 1305 | |||
U70324.1 | - | 11 | 1306 |