Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008077.4 | NP_032103.2 | 17 | 2170 | ||
GenBank mRNA | AK136801.1 | - | 17 | 2155 | ||
AK148497.1 | - | 17 | 2170 | |||
AK158827.1 | - | 16 | 1936 | |||
AK160410.1 | - | 16 | 2079 | |||
AK160868.1 | - | 17 | 2168 | |||
AK163893.1 | - | 3 | 515 | |||
BC027059.1 | - | 17 | 2121 | 5358787 | ||
Y12257.1 | - | 16 | 1937 | |||
Z49976.1 | - | 17 | 2057 |