Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF002701.1 | - | 9 | 1302 | ||
AF079107.1 | - | 8 | 1315 | |||
AF079108.1 | - | 7 | 1231 | |||
AK138512.1 | - | 9 | 2028 | |||
AK149349.1 | - | 9 | 2032 | |||
AK154579.1 | - | 9 | 2040 | |||
BC138561.1 | - | 9 | 1423 | 8861581 |