Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008133.4 | NP_032159.1 | 6 | 1071 | ||
GenBank mRNA | AK075971.1 | - | 6 | 975 | ||
AK089152.1 | - | 6 | 974 | |||
AK150637.1 | - | 6 | 1071 | |||
AK161876.1 | - | 2 | 205 | |||
BC052724.1 | - | 6 | 1045 | 6812055 | ||
BC057347.1 | - | 6 | 1045 | 6812055 | ||
X57024.1 | - | 6 | 976 |