Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008155.4 | NP_032181.2 | 18 | 1795 | ||
GenBank mRNA | AK031081.1 | - | 18 | 1700 | ||
AK076424.1 | - | 16 | 1451 | |||
AK133827.1 | - | 18 | 1717 | |||
AK134890.1 | - | 18 | 1729 | |||
AK137805.1 | - | 18 | 1723 | |||
AK147124.1 | - | 18 | 1758 | |||
AK150341.1 | - | 18 | 1726 | |||
AK152508.1 | - | 13 | 1099 | |||
AK159838.1 | - | 18 | 1699 | |||
AK169681.1 | - | 18 | 1724 | |||
BC086640.1 | - | 18 | 1742 | 6832250 | ||
BC088995.1 | - | 18 | 1702 | 6403790 | ||
L09104.1 | - | 9 | 2934 | |||
M14220.1 | - | 18 | 1724 |