Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163465.1 | NP_001156937.1 | 5 | 543 | ||
NM_008164.2 | NP_032190.2 | 5 | 543 | |||
GenBank mRNA | AK143132.1 | - | 5 | 524 | ||
AK220431.1 | - | 5 | 460 | |||
BC004843.1 | - | 5 | 513 | |||
BC052010.1 | - | 5 | 502 | 5717786 | ||
U43194.1 | - | 5 | 437 |