Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Has3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008217.4 | NP_032243.2 | 4 | 4093 | ||
GenBank mRNA | AK028582.1 | - | 4 | 4037 | ||
U86408.2 | - | 4 | 4099 |