Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V0A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130020.1 | NP_001123492.1 | 2, 3 | 277 | ||
NM_001130021.1 | NP_001123493.1 | 2, 3 | 277 | |||
NM_005177.3 | NP_005168.2 | 2, 3 | 277 | |||
XM_005257459.1 | XP_005257516.1 | 2, 3 | 259 | |||
XM_005257461.1 | XP_005257518.1 | 2, 3 | 259 | |||
XM_005257463.2 | XP_005257520.1 | 2, 3 | 259 | |||
XM_011524908.1 | XP_011523210.1 | 2, 3 | 259 | |||
XM_017024766.1 | XP_016880255.1 | 2, 3 | 259 | |||
XM_017024767.1 | XP_016880256.1 | 2, 3 | 243 | |||
XM_017024768.1 | XP_016880257.1 | 2, 3 | 243 | |||
XM_017024769.1 | XP_016880258.1 | 2, 3 | 243 | |||
XM_017024770.1 | XP_016880259.1 | 2, 3 | 243 | |||
XM_017024771.1 | XP_016880260.1 | 2, 3 | 243 | |||
XM_017024772.1 | XP_016880261.1 | 1, 2 | 62 | |||
XM_017024773.1 | XP_016880262.1 | 1, 2 | 62 | |||
XM_017024774.1 | XP_016880263.1 | 2, 3 | 222 | |||
GenBank mRNA | AK223554.1 | - | 2, 3 | 244 | ||
AK294512.1 | - | 2, 3 | 233 | |||
AK295682.1 | - | 2, 3 | 180 | |||
AK299784.1 | - | 2, 3 | 245 | |||
AK316305.1 | - | 2, 3 | 247 | |||
BC032398.1 | - | 2, 3 | 209 | 5195776 | ||
BX648978.1 | - | 2, 3 | 239 | |||
L78933.1 | - | 2, 3 | 279 | |||
Z71460.1 | - | 2, 3 | 279 |