Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008264.1 | NP_032290.1 | 2 | 1155 | ||
GenBank mRNA | BC100731.1 | - | 3 | 624 | 40045367 | |
BC100732.1 | - | 3 | 621 | 40045368 | ||
BC100733.1 | - | 3 | 621 | 40045370 |