Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp90ab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008302.3 | NP_032328.2 | 3 | 282 | ||
GenBank mRNA | AF465639.1 | - | 3 | 282 | ||
AK030768.1 | - | 3 | 288 | |||
AK084097.1 | - | 3 | 290 | |||
AK135366.1 | - | 3 | 290 | |||
AK146205.1 | - | 3 | 293 | |||
AK146809.1 | - | 3 | 254 | |||
AK178129.1 | - | 2 | 67 | |||
AK179913.1 | - | 2 | 68 | |||
AK180968.1 | - | 2 | 67 | |||
AK184233.1 | - | 3 | 234 | |||
AK190429.1 | - | 2 | 67 | |||
AK196204.1 | - | 2 | 67 | |||
AK218770.1 | - | 2 | 67 | |||
BC002167.1 | - | 3 | 273 | |||
BC088985.1 | - | 3 | 276 | 6808699 | ||
M18186.1 | - | 3 | 281 | |||
M36829.1 | - | 3 | 270 |