Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272073.1 | NP_001259002.1 | 6 | 917 | ||
NM_005051.2 | NP_005042.1 | 6 | 950 | |||
NR_073590.1 | 6 | 901 | ||||
XM_017006965.1 | XP_016862454.1 | 6 | 630 | |||
GenBank mRNA | AF130067.1 | - | 2 | 90 | ||
AK056815.1 | - | 6 | 529 | |||
AK222510.1 | - | 6 | 578 | |||
AK290523.1 | - | 6 | 578 | |||
AK293260.1 | - | 5 | 477 | |||
AK297987.1 | - | 5 | 424 | |||
AK300220.1 | - | 5 | 515 | |||
AK301559.1 | - | 6 | 546 | |||
AK302867.1 | - | 6 | 947 | |||
BC000394.2 | - | 6 | 559 | 2821788 | ||
BC001567.1 | - | 6 | 557 | 3452261 | ||
BC001772.1 | - | 2 | 85 | |||
BC016634.1 | - | 5 | 382 | |||
BC029739.1 | - | 6 | 560 | 3857444 | ||
X76013.1 | - | 6 | 559 |