Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290423.1 | NP_001277352.1 | 5 | 682 | ||
NM_008350.4 | NP_032376.1 | 5 | 781 | |||
GenBank mRNA | AK132897.1 | - | 5 | 683 | ||
BC134354.1 | - | 5 | 643 | 40054168 | ||
U03421.1 | - | 5 | 674 |