Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005063.4 | NP_005054.3 | 2 | 678 | ||
GenBank mRNA | AB032261.1 | - | 2 | 568 | ||
AB208982.1 | - | 2 | 435 | |||
AF097514.1 | - | 2 | 423 | |||
AK222862.1 | - | 2 | 395 | |||
AK312312.1 | - | 2 | 460 | |||
BC005807.2 | - | 2 | 365 | |||
BC062303.1 | - | 2 | 564 | 6586153 | ||
BI827092.1 | - | 2 | 680 | |||
S70284.1 | - | 1 | 183 | |||
Y13647.1 | - | 2 | 396 |