Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010149.3 | NP_034279.3 | 3 | 484 | ||
GenBank mRNA | AK010968.1 | - | 2 | 192 | ||
AK139031.1 | - | 3 | 383 | |||
BC003953.1 | - | 3 | 337 | |||
BC046282.1 | - | 3 | 414 | 6438591 | ||
J04843.1 | - | 3 | 366 |