Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110228.1 | NP_001103698.1 | 2 | 182 | ||
NM_001110229.1 | NP_001103699.1 | 3 | 269 | |||
NM_001110230.1 | NP_001103700.1 | 3 | 269 | |||
NM_001110231.1 | NP_001103701.1 | 5 | 244 | |||
NM_001110232.1 | NP_001103702.1 | 4 | 255 | |||
NM_001160292.1 | NP_001153764.1 | 3 | 269 | |||
NM_001160293.1 | NP_001153765.1 | 2 | 182 | |||
NM_010160.2 | NP_034290.2 | 2 | 182 | |||
GenBank mRNA | AF090696.1 | - | 4 | 255 | ||
AF090697.1 | - | 2 | 155 | |||
AK037436.1 | - | 1 | 641 | |||
AK135166.1 | - | 2 | 133 | |||
AK137292.1 | - | 3 | 210 | |||
AK137555.1 | - | 1 | 40 | |||
AK151818.1 | - | 1 | 40 | |||
AK152628.1 | - | 1 | 40 | |||
AK153267.1 | - | 1 | 40 | |||
AK159996.1 | - | 3 | 276 | |||
AK160861.1 | - | 5 | 245 | |||
AK162651.1 | - | 1 | 40 | |||
AK164112.1 | - | 2 | 79 | |||
AK164914.1 | - | 3 | 210 | |||
AK169542.1 | - | 1 | 55 | |||
AK172648.1 | - | 1 | 40 | |||
AK193426.1 | - | 2 | 211 | |||
AK213125.1 | - | 2 | 105 | |||
BC026856.1 | - | 2 | 133 | 4503295 | ||
BY145030.1 | - | 2 | 139 | |||
BY769645.1 | - | 269 | ||||
CF536042.1 | - | 2 | 182 |