Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eya1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010164.2 | NP_034294.2 | 3, 4 | 735 | ||
GenBank mRNA | AK153788.1 | - | 3, 4 | 740 | ||
BC060260.1 | - | 3, 4 | 735 | 6848408 | ||
BC066860.1 | - | 3, 4 | 392 | 30089692 | ||
U61110.1 | - | 2, 3 | 280 |