Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JMJD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114632.1 | NP_001108104.1 | 5 | 619 | ||
GenBank mRNA | AK000550.1 | - | 5 | 725 | ||
AK000663.1 | - | 5 | 593 | |||
AK000814.1 | - | 4 | 526 | |||
AK225503.1 | - | 5 | 593 | |||
AK289833.1 | - | 5 | 619 | |||
AK290051.1 | - | 5 | 588 | |||
BC025290.1 | - | 5 | 581 | |||
BC139899.1 | - | 5 | 598 |
Gene Symbol: JMJD7-PLA2G4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198588.1 | NP_001185517.1 | 5 | 619 | ||
NM_005090.3 | NP_005081.1 | 5 | 619 | |||
GenBank mRNA | AB209150.1 | - | 5 | 609 | ||
AF065215.1 | - | 5 | 594 | |||
AF065216.1 | - | 4 | 670 | |||
AF121908.1 | - | 5 | 586 | |||
DQ523799.1 | - | 5 | 587 | |||
DQ523800.1 | - | 5 | 587 |