Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc22d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077364.1 | NP_001070832.1 | 4 | 921 | ||
NM_010286.4 | NP_034416.3 | 3 | 746 | |||
GenBank mRNA | AF024519.1 | - | 3 | 731 | ||
AF201287.1 | - | 3 | 490 | |||
AF201288.1 | - | 3 | 726 | |||
AF201289.1 | - | 4 | 883 | |||
AK083389.1 | - | 3 | 745 | |||
BC028813.1 | - | 3 | 736 | 5136440 | ||
EU818782.1 | - | 4 | 920 |