Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281969.1 | NP_001268898.1 | 10 | 2081 | ||
NM_010298.6 | NP_034428.2 | 10 | 2131 | |||
GenBank mRNA | AK082578.1 | - | 10 | 2134 | ||
AK083251.1 | - | 9 | 1985 | |||
BC027094.1 | - | 10 | 2085 | |||
BC037605.1 | - | 10 | 2099 | 5361798 | ||
U09399.1 | - | 10 | 2030 |