Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252555.1 | NP_001239484.1 | 10 | 1246 | ||
NM_001252556.1 | NP_001239485.1 | 8 | 875 | |||
NM_010363.4 | NP_034493.1 | 9 | 943 | |||
GenBank mRNA | AB041613.1 | - | 10 | 1246 | ||
AF093418.1 | - | 9 | 892 | |||
AK002398.1 | - | 9 | 900 | |||
AK012796.1 | - | 9 | 944 | |||
AK075927.1 | - | 9 | 897 | |||
BC031777.1 | - | 9 | 932 | 4168439 |