Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hipk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301304.1 | NP_001288233.1 | 4 | 1449 | ||
NM_001301306.1 | NP_001288235.1 | 4 | 1559 | |||
NM_010432.2 | NP_034562.2 | 4 | 1559 | |||
GenBank mRNA | AF071070.1 | - | 4 | 1449 | ||
AF071071.1 | - | 4 | 1447 | |||
AF077658.1 | - | 4 | 1463 | |||
AF170303.1 | - | 4 | 1359 | |||
AF170304.1 | - | 4 | 1359 | |||
AK134408.1 | - | 4 | 1560 | |||
BC145697.1 | - | 4 | 1312 | 40131000 | ||
BC145699.1 | - | 4 | 1312 | 40131002 | ||
DQ279085.1 | - | 2 | 130 | |||
DQ279089.1 | - | 2 | 106 |