Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmg20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163165.1 | NP_001156637.1 | 7, 8 | 762 | ||
NM_001163166.1 | NP_001156638.1 | 7, 8 | 659 | |||
NM_010440.3 | NP_034570.1 | 6, 7 | 846 | |||
GenBank mRNA | AF146224.1 | - | 6, 7 | 829 | ||
AK153816.1 | - | 7, 8 | 660 | |||
AK155652.1 | - | 7, 8 | 664 | |||
AK155890.1 | - | 6, 7 | 722 | |||
AK183154.1 | - | 3, 4 | 239 | |||
AK186994.1 | - | 3, 4 | 239 | |||
AK187890.1 | - | 3, 4 | 159 | |||
AK189436.1 | - | 3, 4 | 239 | |||
AK216551.1 | - | 3, 4 | 239 | |||
AL355735.1 | - | 3, 4 | 221 | |||
BC003924.1 | - | 6, 7 | 734 | 3485302 | ||
BC011334.1 | - | 7, 8 | 695 | 4012515 | ||
BC023436.1 | - | 4, 5 | 322 | |||
BC071217.1 | - | 4, 5 | 1723 |