Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ids | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010498.3 | NP_034628.2 | 3 | 585 | ||
GenBank mRNA | AJ627044.1 | - | 1 | 50 | ||
AK085237.1 | - | 4 | 462 | |||
AK132287.1 | - | 4 | 465 | |||
AK166178.1 | - | 3 | 463 | |||
AK166443.1 | - | 4 | 414 | |||
L07921.1 | - | 3 | 462 |