Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010501.2 | NP_034631.1 | 2 | 1267 | ||
GenBank mRNA | AK077459.1 | - | 2 | 1266 | ||
AK151032.1 | - | 2 | 1267 | |||
AK153226.1 | - | 2 | 1268 | |||
BC003804.1 | - | 2 | 1233 | 3487345 | ||
BC089563.1 | - | 2 | 1265 | 30317428 | ||
CT010266.1 | - | 1 | 1196 | |||
L32974.1 | - | 2 | 1248 |