Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifnar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010509.2 | NP_034639.2 | 8 | 1131 | ||
GenBank mRNA | AF013274.1 | - | 8 | 909 | ||
AK139169.1 | - | 8 | 1111 | |||
AK150386.1 | - | 8 | 1065 | |||
AK150597.1 | - | 8 | 1044 | |||
AK151474.1 | - | 8 | 1059 | |||
AK151493.1 | - | 8 | 1044 | |||
AK151777.1 | - | 8 | 1059 | |||
AK152878.1 | - | 8 | 1065 | |||
AK153317.1 | - | 8 | 1065 | |||
BC071225.1 | - | 7 | 829 | 4187603 | ||
Y09865.1 | - | 8 | 1021 |