Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252662.1 | NP_001239591.1 | 16 | 2002 | ||
NM_001252663.1 | NP_001239592.1 | 15 | 1936 | |||
NM_001252664.1 | NP_001239593.1 | 15 | 1829 | |||
NM_001252665.1 | NP_001239594.1 | 14 | 1662 | |||
NM_001252666.1 | NP_001239595.1 | 13 | 1695 | |||
NM_013514.4 | NP_038542.1 | 16 | 1877 | |||
GenBank mRNA | AF079846.3 | - | 14 | 1606 | ||
AF155547.1 | - | 15 | 1672 | |||
AK143840.1 | - | 16 | 1996 | |||
AK149301.1 | - | 15 | 1639 | |||
AK155927.1 | - | 15 | 1931 | |||
AK158744.1 | - | 15 | 1760 | |||
AK162866.1 | - | 14 | 1614 | |||
AK165204.1 | - | 1995 | ||||
AK165294.1 | - | 15 | 1928 | |||
AK165709.1 | - | 1 | 275 | |||
BC016897.1 | - | 16 | 1877 | 4240747 | ||
BC037021.1 | - | 13 | 1682 | 5366040 |