Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il2rg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK143669.1 | - | 4, 5 | 624 | ||
AK153873.1 | - | 4, 5 | 639 | |||
BC014720.1 | - | 4, 5 | 618 | 4924951 | ||
D13565.1 | - | 4, 5 | 616 | |||
D13821.1 | - | 4, 5 | 611 | |||
KC815469.1 | - | 4, 5 | 614 | |||
L20048.1 | - | 4, 5 | 628 | |||
X75337.1 | - | 4, 5 | 641 |