Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013818.2 | NP_038846.2 | 8 | 1364 | ||
GenBank mRNA | AK004612.1 | - | 8 | 1365 | ||
AK147717.1 | - | 8 | 1366 | |||
AK150068.1 | - | 8 | 1371 | |||
AK150185.1 | - | 8 | 1371 | |||
AK158342.1 | - | 8 | 1374 | |||
AK170091.1 | - | 8 | 1366 | |||
BC037129.1 | - | 6 | 906 | |||
BC046228.1 | - | 8 | 1364 | 5708774 | ||
U87965.1 | - | 8 | 1251 |