Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170981.1 | NP_001164452.1 | 9 | 1126 | ||
NM_001170982.1 | NP_001164453.1 | 8 | 1100 | |||
NM_001170983.1 | NP_001164454.1 | 9 | 1105 | |||
NM_001170984.1 | NP_001164455.1 | 9 | 1102 | |||
NM_016884.3 | NP_058580.1 | 9 | 1165 | |||
GenBank mRNA | AF095257.1 | - | 9 | 1119 | ||
AK010845.1 | - | 9 | 1080 | |||
AK011336.1 | - | 9 | 1098 | |||
AK012633.2 | - | 9 | 1088 | |||
AK019958.1 | - | 9 | 2280 | |||
AK088245.1 | - | 9 | 1142 | |||
AK088678.1 | - | 9 | 1116 | |||
AK089061.1 | - | 9 | 1106 | |||
AK150399.1 | - | 9 | 1139 | |||
AK151446.1 | - | 9 | 1141 | |||
AK153053.1 | - | 9 | 1141 | |||
AK166590.1 | - | 9 | 1083 | |||
AK168802.1 | - | 9 | 1100 | |||
AK169287.1 | - | 10 | 1022 | |||
AK219753.1 | - | 3 | 297 | |||
BC004706.1 | - | 9 | 1064 | 3499283 | ||
BC095922.1 | - | 8 | 1041 | 4988965 | ||
CK620126.1 | - | 4 | 461 |